Assay Details
Target Gene Details
Entrez Gene ID: | 14824 |
Gene Name: | granulin |
Gene Aliases: |
Pgrn, epithelin |
Location: |
Chr.11:102430322-102436809 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 14824 |
Gene Name: | granulin |
Gene Aliases: |
Pgrn, epithelin |
Location: |
Chr.11:102430322-102436809 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |