Assay Details
Target Gene Details
Entrez Gene ID: | 66213 |
Gene Name: | mediator complex subunit 7 |
Gene Aliases: |
1110063B05Rik, Crsp33, Crsp9 |
Location: |
Chr.11:46436933-46442721 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med7 | NM_001104530.1 | 2 | 186 | NP_001098000.1 |
| NM_001104556.1 | NP_001098026.1 | |||
| NM_001104557.1 | NP_001098027.1 | |||
| NM_025426.3 | 2 | 354 | NP_079702.3 | |
| AK004354.1 | 2 | 319 | BAB23272.1 | |
| AK010357.1 | 2 | 182 | BAB26877.1 | |
| AK012799.1 | 2 | 334 | BAB28478.1 | |
| AK144918.1 | BAE26135.1 | |||
| AK147739.1 | 2 | 344 | BAE28106.1 | |
| AK167407.1 | 2 | 314 | BAE39496.1 | |
| AK185444.1 | ||||
| BC011333.1 | 2 | 341 | AAH11333.1 | |
| BE333513.1 | ||||
| BY135693.1 | 2 | 184 | ||
| C88719.1 | ||||
| CJ145922.1 | 2 | 356 | ||
| CK030901.1 | 2 | 186 |
More Information
Set Membership: |
|

Genomic Map