Assay Details
Target Gene Details
Entrez Gene ID: | 17884 |
Gene Name: | myosin, heavy polypeptide 4, skeletal muscle |
Gene Aliases: |
AI506973, MHC2B, MM, MYH-2B, Minimsc, Minmus, MyHC-IIb, Myhsf |
Location: |
Chr.11:67237812-67260447 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 33 - Intron 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myh4 | NM_010855.3 | NP_034985.2 | ||
| AJ278733.1 | CAC00537.1 | |||
| BC141362.1 |

Genomic Map