Assay Details
Target Gene Details
Entrez Gene ID: | 22780 |
Gene Name: | IKAROS family zinc finger 3 |
Gene Aliases: |
5830411O07Rik, Aiolos, Zfpn1a3, Znfn1a3 |
Location: |
Chr.11:98464610-98546380 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikzf3 | NM_011771.1 | NP_035901.1 | ||
| AF001293.1 | AAB58795.1 | |||
| AI549961.1 | ||||
| AK036657.1 | ||||
| AK157177.1 | ||||
| AK169802.1 | 1 | 675 |
More Information
Set Membership: |
|

Genomic Map