Assay Details
Target Gene Details
Entrez Gene ID: | 16407 |
Gene Name: | integrin alpha E, epithelial-associated |
Gene Aliases: |
A530055J10, CD103, aM290, alpha-E1, alpha-M290 |
Location: |
Chr.11:73090595-73147449 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgae | NM_008399.2 | 3 | 235 | NP_032425.2 |
| NM_172944.2 | 3 | 235 | NP_766532.1 | |
| AK040983.1 | 3 | 236 | BAC30769.1 | |
| AK154021.1 | 3 | 236 | BAE32321.1 | |
| BC150690.1 | 3 | 235 | ||
| DQ386608.1 | 3 | 235 | ||
| U12236.1 | 3 | 259 | AAC52142.1 |
More Information
Set Membership: |
|

Genomic Map