Assay Details
Target Gene Details
Entrez Gene ID: | 26936 |
Gene Name: | myosin phosphatase Rho interacting protein |
Gene Aliases: |
9530046C02, AA536749, AI647711, C76423, RIP3, Rhoip3, mKIAA0864, p116Rip |
Location: |
Chr.11:59662473-59780860 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mprip | NM_012027.2 | 16 | 2606 | NP_036157.2 |
| NM_201245.3 | 16 | 2606 | NP_957697.1 | |
| AB093269.1 | 4 | 4431 | BAC41453.1 | |
| AK035435.1 | 10 | 1896 | BAC29064.1 | |
| AK154807.1 | 17 | 2613 | BAE32843.1 | |
| AK155630.1 | 16 | 2387 | BAE33355.1 | |
| AK158296.1 | 16 | 2595 | BAE34450.1 | |
| AK162530.1 | 14 | 1952 | BAE36957.1 | |
| AK165710.1 | 2 | 765 | ||
| AK170849.1 | 16 | 2590 | BAE42072.1 | |
| AK171089.1 | 16 | 2595 | BAE42242.1 | |
| BC049803.1 | 16 | 2132 | AAH49803.1 | |
| BC063067.1 | 16 | 2569 | AAH63067.1 | |
| U73200.1 | 16 | 2466 | AAB18198.1 |
More Information
Set Membership: |
|

Genomic Map