Assay Details
Target Gene Details
Entrez Gene ID: | 16897 |
Gene Name: | lethal giant larvae homolog 1 (Drosophila) |
Gene Aliases: |
AI325176, Lgl1, Llglh, Mgl1 |
Location: |
Chr.11:60699670-60714188 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Llgl1 | NM_001159404.1 | NP_001152876.1 | ||
| NM_001159405.1 | NP_001152877.1 | |||
| NM_008502.2 | NP_032528.1 | |||
| AK159412.1 | BAE35062.1 | |||
| BC050913.1 | AAH50913.1 | |||
| BC055399.1 | AAH55399.1 | |||
| D16141.1 | BAA03712.1 |
More Information
Set Membership: |
|

Genomic Map