Assay Details
Target Gene Details
Entrez Gene ID: | 76293 |
Gene Name: | microfibrillar-associated protein 4 |
Gene Aliases: |
1110007F23Rik |
Location: |
Chr.11:61485378-61488706 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfap4 | NM_029568.2 | 5 | 505 | NP_083844.1 |
| AK003537.1 | 5 | 505 | BAB22844.1 | |
| AK075659.1 | 5 | 567 | ||
| BC022666.1 | 5 | 497 | AAH22666.1 |
More Information
Set Membership: |
|

Genomic Map