Assay Details
Target Gene Details
Entrez Gene ID: | 18015 |
Gene Name: | neurofibromatosis 1 |
Gene Aliases: |
AW494271, E030030H24Rik, Nf-1 |
Location: |
Chr.11:79335988-79581609 on Build GRCm38 |
Assay Gene Location: | Within Intron 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nf1 | NM_010897.2 | NP_035027.1 | ||
| L10370.1 | AAA68132.1 |
Target Gene Details
Entrez Gene ID: | 18377 |
Gene Name: | oligodendrocyte myelin glycoprotein |
Gene Aliases: |
- |
Location: |
Chr.11:79500982-79504082 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Omg | NM_019409.2 | NP_062282.2 | ||
| AK136910.1 | BAE23166.1 | |||
| BC024757.1 | AAH24757.1 | |||
| BY721452.1 |
More Information
Set Membership: |
|

Genomic Map