Assay Details
Target Gene Details
Entrez Gene ID: | 70834 |
Gene Name: | sperm associated antigen 9 |
Gene Aliases: |
3110018C07Rik, 4733401I23Rik, 4831406C20Rik, AW552012, JLP, JSAP2, JSAP2a, Jip4, Mapk8ip4, syd1 |
Location: |
Chr.11:93996058-94126087 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spag9 | NM_001025428.1 | 15 | 1804 | NP_001020599.1 |
| NM_001025429.1 | 15 | 1801 | NP_001020600.1 | |
| NM_001025430.1 | 14 | 1789 | NP_001020601.1 | |
| NM_001199203.1 | 18 | 2280 | NP_001186132.1 | |
| NM_001199204.1 | 17 | 2238 | NP_001186133.1 | |
| NM_001199205.1 | 14 | 1789 | NP_001186134.1 | |
| NM_027569.2 | 17 | 2238 | NP_081845.2 | |
| AB047782.1 | 18 | 2200 | BAD93176.1 | |
| AF327451.1 | 17 | 2220 | AAN61564.1 | |
| AK014638.1 | 9 | 1298 | ||
| AK049161.1 | 8 | 801 | BAC33577.1 | |
| AK147431.1 | 14 | 1846 | BAE27907.1 | |
| AK147537.1 | 14 | 1865 | BAE27980.1 | |
| AK172961.1 | 15 | 1783 | BAD32239.1 | |
| AY823270.1 | 17 | 1782 | AAX19462.1 | |
| BC060100.1 | 14 | 1723 | AAH60100.1 | |
| BC060506.1 | 2 | 122 | AAH60506.1 | |
| BC094670.1 | 14 | 1717 | AAH94670.1 | |
| FJ042496.1 | 17 | 2115 |
More Information
Set Membership: |
|

Genomic Map