Assay Details
Target Gene Details
Entrez Gene ID: | 320207 |
Gene Name: | phosphoinositide-3-kinase, regulatory subunit 5, p101 |
Gene Aliases: |
AV230647, F730038I15Rik |
Location: |
Chr.11:68432125-68497849 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3r5 | NM_177320.2 | 12 | 1965 | NP_796294.2 |
| AK089483.1 | 12 | 1970 | BAC40901.2 | |
| AK149847.1 | 12 | 1933 | BAE29119.1 | |
| AK154643.1 | 12 | 1944 | ||
| AK161050.1 | 3 | 873 | BAE36171.1 | |
| AK171279.1 | 11 | 2090 | ||
| AY156924.1 | 11 | 1814 | AAN84787.1 | |
| BC128076.1 | 11 | 1814 | ||
| BC128077.1 | 11 | 1814 |
More Information
Set Membership: |
|

Genomic Map