Assay Details
Target Gene Details
Entrez Gene ID: | 380714 |
Gene Name: | rabphilin 3A-like (without C2 domains) |
Gene Aliases: |
Gm1753, Noc2 |
Location: |
Chr.11:75830992-75967250 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rph3al | NM_001291159.1 | NP_001278088.1 | ||
| NM_029548.4 | NP_083824.1 | |||
| AA616014.1 | ||||
| AB112925.1 | BAD07030.1 | |||
| AB158403.1 | BAD18853.1 | |||
| AI594476.1 | ||||
| AK018335.1 | BAB31169.1 | |||
| BC127960.1 | ||||
| DV651301.1 | ||||
| DV660756.1 |

Genomic Map