Assay Details
Target Gene Details
Entrez Gene ID: | 69215 |
Gene Name: | spermidine/spermine N1-acetyl transferase 2 |
Gene Aliases: |
2610016A03Rik, SSAT-2, SSAT2 |
Location: |
Chr.11:69621993-69624034 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sat2 | NM_026991.2 | 1 | 119 | NP_081267.1 |
| AK011406.1 | ||||
| AK161230.1 | 1 | 177 | BAE36254.1 | |
| BC061227.1 | 1 | 119 | AAH61227.1 |
More Information
Set Membership: |
|

Genomic Map