Assay Details
Target Gene Details
Entrez Gene ID: | 320207 |
Gene Name: | phosphoinositide-3-kinase, regulatory subunit 5, p101 |
Gene Aliases: |
AV230647, F730038I15Rik |
Location: |
Chr.11:68432125-68497849 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3r5 | NM_177320.2 | 7 | 758 | NP_796294.2 |
| AK029201.1 | 5 | 426 | ||
| AK089483.1 | 7 | 763 | BAC40901.2 | |
| AK149847.1 | 7 | 726 | BAE29119.1 | |
| AK154643.1 | 7 | 736 | ||
| AK171279.1 | 7 | 759 | ||
| AY156924.1 | 6 | 607 | AAN84787.1 | |
| BC128076.1 | 6 | 607 | ||
| BC128077.1 | 6 | 607 |
More Information
Set Membership: |
|

Genomic Map