Assay Details
Target Gene Details
Entrez Gene ID: | 68662 |
Gene Name: | secretoglobin, family 3A, member 1 |
Gene Aliases: |
HIN-1, LuLeu2, Pnsp2, UGRP2, Utgrp2 |
Location: |
Chr.11:49663595-49665118 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scgb3a1 | NM_054037.2 | NP_473378.1 | ||
| NM_170727.2 | NP_733923.1 | |||
| AF313456.1 | AAL26216.1 | |||
| AF313457.1 | AAN62328.1 | |||
| AF436840.1 | AAQ04482.1 | |||
| AK003988.1 | ||||
| AV578532.1 | ||||
| BC086910.1 | AAH86910.1 |

Genomic Map