Assay Details
Target Gene Details
Entrez Gene ID: | 68662 |
Gene Name: | secretoglobin, family 3A, member 1 |
Gene Aliases: |
HIN-1, LuLeu2, Pnsp2, UGRP2, Utgrp2 |
Location: |
Chr.11:49663595-49665118 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scgb3a1 | NM_054037.2 | 2 | 202 | NP_473378.1 |
| NM_170727.2 | 3 | 307 | NP_733923.1 | |
| AF313456.1 | 2 | 187 | AAL26216.1 | |
| AF313457.1 | 3 | 292 | AAN62328.1 | |
| AF436840.1 | 2 | 171 | AAQ04482.1 | |
| AK003988.1 | 2 | 189 | ||
| AV578532.1 | 2 | 202 | ||
| BC086910.1 | 2 | 184 | AAH86910.1 |

Genomic Map