Assay Details
Target Gene Details
Entrez Gene ID: | 53860 |
Gene Name: | septin 9 |
Gene Aliases: |
MSF1, Msf, PNUTL4, Sint1 |
Location: |
Chr.11:117199661-117362325 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept9 | NM_001113486.1 | NP_001106958.1 | ||
| AK141312.1 | BAE24646.1 | |||
| BC046524.1 | AAH46524.1 |
More Information
Set Membership: |
|

Genomic Map