Assay Details
Target Gene Details
Entrez Gene ID: | 217166 |
Gene Name: | nuclear receptor subfamily 1, group D, member 1 |
Gene Aliases: |
A530070C09Rik, R75201 |
Location: |
Chr.11:98767932-98775377 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr1d1 | NM_145434.4 | NP_663409.2 | ||
| AF291821.1 | AAG01345.1 | |||
| AK041047.1 | ||||
| AK053029.1 | 1 | 2446 | ||
| AK135640.1 | ||||
| AK137398.1 | BAE23342.1 | |||
| AK137582.1 | BAE23418.1 | |||
| AK146430.1 | BAE27164.1 | |||
| AK154931.1 | BAE32932.1 | |||
| AK155597.1 | BAE33339.1 | |||
| BC008989.1 | AAH08989.1 |
More Information
Set Membership: |
|

Genomic Map