Assay Details
Target Gene Details
Entrez Gene ID: | 67245 |
Gene Name: | pellino 1 |
Gene Aliases: |
2810468L03Rik, A930031K15Rik, AA409794, AI586297, D11Ertd676e |
Location: |
Chr.11:21091324-21150327 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Peli1 | NM_023324.2 | 5 | 831 | NP_075813.2 |
| AF302503.1 | 4 | 1053 | AAG15391.1 | |
| AK013388.1 | ||||
| AK029586.1 | 4 | 2591 | ||
| AK151660.1 | 5 | 832 | BAE30588.1 | |
| AK152900.1 | 5 | 832 | BAE31581.1 | |
| AK152997.1 | 5 | 831 | ||
| AK153372.1 | 5 | 832 | BAE31940.1 | |
| AK167525.1 | 5 | 832 | BAE39595.1 | |
| BC016515.1 | 4 | 430 | AAH16515.1 |
More Information
Set Membership: |
|

Genomic Map