Assay Details
Target Gene Details
Entrez Gene ID: | 237987 |
Gene Name: | otopetrin 2 |
Gene Aliases: |
4732464P15 |
Location: |
Chr.11:115307163-115332306 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Otop2 | NM_172801.2 | NP_766389.2 | ||
| AB087504.1 | BAC67684.1 | |||
| AK028866.1 | BAC26159.1 |
More Information
Set Membership: |
|

Genomic Map