Assay Details
Target Gene Details
Entrez Gene ID: | 217212 |
Gene Name: | peptide YY |
Gene Aliases: |
- |
Location: |
Chr.11:102106676-102107859 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pyy | NM_145435.1 | 4 | 364 | NP_663410.1 |
| AK050518.1 | 4 | 421 | BAE20675.1 | |
| BC010821.1 | 4 | 364 | AAH10821.1 |
More Information
Set Membership: |
|

Genomic Map