Assay Details
Target Gene Details
Entrez Gene ID: | 53860 |
Gene Name: | septin 9 |
Gene Aliases: |
MSF1, Msf, PNUTL4, Sint1 |
Location: |
Chr.11:117199661-117362325 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept9 | NM_001113486.1 | NP_001106958.1 | ||
| NM_001113487.1 | NP_001106959.1 | |||
| NM_001113488.1 | NP_001106960.1 | |||
| NM_017380.2 | NP_059076.1 | |||
| AJ250723.1 | CAB59833.1 | |||
| AK031757.1 | BAC27538.1 | |||
| AK122415.2 | BAC65697.2 | |||
| AK141312.1 | BAE24646.1 | |||
| AK164581.1 | 8 | 929 | ||
| BC046524.1 | AAH46524.1 |
More Information
Set Membership: |
|

Genomic Map