Assay Details
Target Gene Details
Entrez Gene ID: | 78473 |
Gene Name: | src family associated phosphoprotein 1 |
Gene Aliases: |
1700091G21Rik, Scap1, Skap-55 |
Location: |
Chr.11:96464576-96759622 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skap1 | NM_001033186.3 | NP_001028358.1 | ||
| NM_001177898.1 | NP_001171369.1 | |||
| NM_001177899.1 | NP_001171370.1 | |||
| AK018920.1 | ||||
| AK137942.1 | BAE23518.1 | |||
| AK138030.1 | BAE23537.1 | |||
| AK169825.1 | BAE41394.1 | |||
| BC145402.1 | ||||
| BC150764.1 | ||||
| BC171961.1 |
More Information
Set Membership: |
|

Genomic Map