Assay Details
Target Gene Details
Entrez Gene ID: | 78473 |
Gene Name: | src family associated phosphoprotein 1 |
Gene Aliases: |
1700091G21Rik, Scap1, Skap-55 |
Location: |
Chr.11:96464576-96759622 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skap1 | NM_001033186.3 | NP_001028358.1 | ||
| NM_001177898.1 | NP_001171369.1 | |||
| NM_001177899.1 | NP_001171370.1 | |||
| AK040068.1 | BAE20569.1 | |||
| AK041917.1 | BAE20612.1 | |||
| AK137792.1 | BAE23497.1 | |||
| AK137942.1 | BAE23518.1 | |||
| AK138030.1 | BAE23537.1 | |||
| AK153578.1 | BAE32106.1 | |||
| AK162116.1 | ||||
| AK169685.1 | ||||
| AK169825.1 | BAE41394.1 | |||
| BC145402.1 | ||||
| BC150764.1 | ||||
| BC171961.1 | ||||
| BY723539.1 |
More Information
Set Membership: |
|

Genomic Map