Assay Details
Target Gene Details
Entrez Gene ID: | 18816 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade F, member 2 |
Gene Aliases: |
AI747498, Pli, Serpimf2 |
Location: |
Chr.11:75431736-75439501 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinf2 | NM_008878.2 | NP_032904.1 | ||
| AK077563.1 | ||||
| AK090208.1 | ||||
| BC026756.1 | AAH26756.1 | |||
| GQ896353.1 | ||||
| GQ896354.1 | ||||
| Z36774.1 | CAA85350.1 |
More Information
Set Membership: |
|

Genomic Map