Assay Details
Target Gene Details
Entrez Gene ID: | 18626 |
Gene Name: | period circadian clock 1 |
Gene Aliases: |
Per, m-rigui, mPer1 |
Location: |
Chr.11:69095140-69109965 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Per1 | NM_001159367.1 | NP_001152839.1 | ||
| NM_011065.4 | NP_035195.2 | |||
| AB002108.1 | BAA22634.1 | |||
| AF022992.1 | AAC53355.1 | |||
| AK081813.1 | BAC38339.1 | |||
| AK148202.1 | BAE28412.1 | |||
| AK154900.1 | BAE32911.1 | |||
| AK172958.1 | BAD32236.1 | |||
| BC039768.1 | AAH39768.1 | |||
| BC091645.1 | AAH91645.1 |
More Information
Set Membership: |
|

Genomic Map