Assay Details
Target Gene Details
Entrez Gene ID: | 72508 |
Gene Name: | ribosomal protein S6 kinase, polypeptide 1 |
Gene Aliases: |
2610318I15Rik, 4732464A07Rik, 70kDa, AA959758, AI256796, AI314060, S6K1, p70/85s6k, p70s6k |
Location: |
Chr.11:86498869-86544807 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps6kb1 | NM_001114334.1 | NP_001107806.1 | ||
| NM_028259.4 | NP_082535.1 | |||
| AK032724.1 | BAC28000.1 | |||
| AK085687.1 | BAC39506.1 | |||
| AK135716.1 | BAE22625.1 | |||
| AK163258.1 | ||||
| AK178169.1 | ||||
| BC038491.1 | AAH38491.1 | |||
| BC071244.1 | AAH71244.1 |
More Information
Set Membership: |
|

Genomic Map