Assay Details
Target Gene Details
Entrez Gene ID: | 50776 |
Gene Name: | polymerase (DNA directed), gamma 2, accessory subunit |
Gene Aliases: |
- |
Location: |
Chr.11:106754821-106780010 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polg2 | NM_015810.2 | NP_056625.2 | ||
| NR_027785.1 | ||||
| AF177202.1 | AAD56641.1 | |||
| AK041591.1 | BAE20595.1 | |||
| AK042777.1 | ||||
| AK164730.1 | ||||
| AK169977.1 | ||||
| BC119131.1 |
More Information
Set Membership: |
|

Genomic Map