Assay Details
Target Gene Details
Entrez Gene ID: | 55963 |
Gene Name: | solute carrier family 1 (glutamate/neutral amino acid transporter), member 4 |
Gene Aliases: |
ASCT-1, ASCT1, AW045657, SATT |
Location: |
Chr.11:20302180-20332713 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc1a4 | NM_018861.3 | NP_061349.3 | ||
| AF246129.1 | AAG02179.1 | |||
| AK044280.1 | BAC31852.1 | |||
| AK136900.1 | BAE23162.1 | |||
| AK139248.1 | BAE23932.1 | |||
| AK140866.1 | BAE24502.1 | |||
| AK148075.1 | BAE28327.1 | |||
| BC043483.1 | AAH43483.1 | |||
| BC052733.1 | AAH52733.1 | |||
| U75215.1 | AAB71740.1 |
More Information
Set Membership: |
|

Genomic Map