Assay Details
Target Gene Details
Entrez Gene ID: | 170758 |
Gene Name: | RAS-related C3 botulinum substrate 3 |
Gene Aliases: |
Rac1B |
Location: |
Chr.11:120721468-120723969 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rac3 | NM_133223.4 | NP_573486.1 | ||
| AB040819.1 | BAB40573.1 | |||
| AK089930.1 | BAC41001.1 | |||
| AK150203.1 | ||||
| AY974601.1 | ||||
| BC025842.1 | AAH25842.1 | |||
| BC117028.1 | ||||
| BC119041.1 |
More Information
Set Membership: |
|

Genomic Map