Assay Details
Target Gene Details
Entrez Gene ID: | 74136 |
Gene Name: | SEC14-like 1 (S. cerevisiae) |
Gene Aliases: |
1200017E04Rik, 2810012L19Rik, Naa-35 |
Location: |
Chr.11:117115172-117159268 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec14l1 | NM_001166506.1 | NP_001159978.1 | ||
| NM_001166507.1 | NP_001159979.1 | |||
| NM_028777.3 | NP_083053.2 | |||
| AK004818.1 | BAB23589.1 | |||
| AK047396.1 | BAC33044.1 | |||
| AK172947.1 | BAD32225.1 | |||
| AK213489.1 | ||||
| BC005766.1 | AAH05766.1 |

Genomic Map