Assay Details
Target Gene Details
Entrez Gene ID: | 20020 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide A |
Gene Aliases: |
220kDa, Rpb1, Rpo2-1 |
Location: |
Chr.11:69733997-69758633 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2a | NM_001291068.1 | 4 | 908 | NP_001277997.1 |
| AK154270.1 | 4 | 910 | ||
| AK154389.1 | 4 | 910 | ||
| AK155391.1 | 4 | 910 | ||
| U37500.1 | 4 | 567 | AAB58418.1 |
More Information
Set Membership: |
|

Genomic Map