Assay Details
Target Gene Details
Entrez Gene ID: | 103554 |
Gene Name: | proteasome (prosome, macropain) activator subunit 4 |
Gene Aliases: |
AA409398, AU041366, TEMO, mKIAA0077 |
Location: |
Chr.11:30771328-30881150 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psme4 | NM_134013.3 | 11 | 1613 | NP_598774.2 |
| AF296169.1 | 11 | 1324 | AAG09060.1 | |
| BC141381.1 | 11 | 1326 | ||
| CV557303.1 | 4 | 482 |

Genomic Map