Assay Details
Target Gene Details
Entrez Gene ID: | 216578 |
Gene Name: | poly(A) polymerase gamma |
Gene Aliases: |
9630006B20Rik |
Location: |
Chr.11:23860145-23895296 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Papolg | NM_172555.2 | NP_766143.2 | ||
| AK035810.1 | BAC29194.1 | |||
| AK042629.1 | BAC31312.1 | |||
| AK139065.1 | BAE23879.1 | |||
| BC059271.1 | AAH59271.1 | |||
| DQ103505.1 | AAZ80292.1 |
More Information
Set Membership: |
|

Genomic Map