Assay Details
Target Gene Details
Entrez Gene ID: | 268373 |
Gene Name: | peptidylprolyl isomerase A |
Gene Aliases: |
2700098C05, Cphn, CyP-18, CypA |
Location: |
Chr.11:6415870-6419810 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppia | NM_008907.1 | NP_032933.1 | ||
| AK002236.1 | BAB21954.1 | |||
| AK007981.1 | BAB25387.1 | |||
| AK010649.1 | BAB27089.1 | |||
| AK012491.1 | BAB28276.1 | |||
| AK012531.1 | BAB28300.1 | |||
| AK012541.1 | ||||
| AK012663.1 | BAB28392.1 | |||
| AK028210.1 | BAC25817.1 | |||
| AK131942.1 | BAE20888.1 | |||
| AK136788.1 | BAE23129.1 | |||
| AK151347.1 | BAE30323.1 | |||
| AK151583.1 | BAE30523.1 | |||
| AK153030.1 | BAE31662.1 | |||
| AK160631.1 | BAE35927.1 | |||
| AK160780.1 | BAE36004.1 | |||
| AK168035.1 | BAE40018.1 | |||
| AK169813.1 | BAE41385.1 | |||
| BC083076.1 | AAH83076.1 | |||
| BC087928.1 | AAH87928.1 | |||
| BC094272.1 | AAH94272.1 | |||
| BC094632.1 | AAH94632.1 | |||
| BC099478.1 | AAH99478.1 | |||
| DQ985731.1 | ||||
| X52803.1 | CAA36989.1 |
More Information
Set Membership: |
|

Genomic Map