Assay Details
Target Gene Details
Entrez Gene ID: | 320472 |
Gene Name: | protein phosphatase 1E (PP2C domain containing) |
Gene Aliases: |
AW049266, B930008A12Rik, POPX1, PP2CH, mKIAA1072 |
Location: |
Chr.11:87226906-87358994 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppm1e | NM_177167.4 | NP_796141.2 | ||
| AK036583.1 | BAC29490.1 | |||
| AK122434.1 | BAC65716.1 | |||
| BC138900.1 | ||||
| BC138903.1 |
Target Gene Details
Entrez Gene ID: | 68729 |
Gene Name: | tripartite motif-containing 37 |
Gene Aliases: |
1110032A10Rik, 2810004E07Rik, AI848587, AU043018, MUL, TEF3, mKIAA0898 |
Location: |
Chr.11:87127051-87250874 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim37 |
More Information
Set Membership: |
|

Genomic Map