Assay Details
Target Gene Details
Entrez Gene ID: | 19049 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 1B |
Gene Aliases: |
AU040756, DARPP-32, Darpp32 |
Location: |
Chr.11:98348738-98357795 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r1b | NM_144828.1 | NP_659077.1 | ||
| AB221544.1 | ||||
| AY640620.1 | AAT66920.1 | |||
| AY640621.1 | 1 | 554 | AAT66921.1 | |
| BC011122.1 | AAH11122.1 | |||
| BC026568.1 | AAH26568.1 | |||
| BC031129.1 | AAH31129.1 |
More Information
Set Membership: |
|

Genomic Map