Assay Details
Target Gene Details
Entrez Gene ID: | 192157 |
Gene Name: | suppressor of cytokine signaling 7 |
Gene Aliases: |
2310063P06Rik, C85125, Nap4 |
Location: |
Chr.11:97362296-97398542 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Socs7 | NM_138657.3 | NP_619598.1 | ||
| AF424814.1 | AAL60516.1 | |||
| AK147580.1 | BAE28006.1 | |||
| AK147650.1 | BAE28049.1 |
More Information
Set Membership: |
|

Genomic Map