Assay Details
Target Gene Details
Entrez Gene ID: | 100041311 |
Gene Name: | predicted gene 11978 |
Gene Aliases: |
OTTMUSG00000005125, OTTMUSG00000005127 |
Location: |
Chr.11:6650148-6660236 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm11978 | NR_028586.1 | |||
| AK132798.1 |
Target Gene Details
Entrez Gene ID: | 56089 |
Gene Name: | receptor (calcitonin) activity modifying protein 3 |
Gene Aliases: |
AI850306 |
Location: |
Chr.11:6658533-6677476 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ramp3 | NM_019511.3 | NP_062384.1 | ||
| AF146524.1 | AAD35020.1 | |||
| AF209907.1 | AAF21039.1 | |||
| AJ250491.1 | CAB59513.1 | |||
| AK005513.1 | ||||
| BC024765.1 | AAH24765.1 |
More Information
Set Membership: |
|

Genomic Map