Assay Details
Target Gene Details
Entrez Gene ID: | 56089 |
Gene Name: | receptor (calcitonin) activity modifying protein 3 |
Gene Aliases: |
AI850306 |
Location: |
Chr.11:6658533-6677476 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ramp3 | NM_019511.3 | 2 | 162 | NP_062384.1 |
| AF146524.1 | 2 | 163 | AAD35020.1 | |
| AF209907.1 | 2 | 122 | AAF21039.1 | |
| AJ250491.1 | 2 | 163 | CAB59513.1 | |
| AK005513.1 | 3 | 278 | ||
| BC024765.1 | 2 | 158 | AAH24765.1 |
More Information
Set Membership: |
|

Genomic Map