Assay Details
Target Gene Details
Entrez Gene ID: | 68275 |
Gene Name: | replication protein A1 |
Gene Aliases: |
5031405K23Rik, 70kDa, AA589576, AW557552, RF-A, RP-A, Rpa |
Location: |
Chr.11:75300259-75348383 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpa1 | NM_001164223.1 | 12 | 1245 | NP_001157695.1 |
| NM_026653.2 | 11 | 1182 | NP_080929.1 | |
| AK019864.1 | 5 | 583 | ||
| AK035084.1 | 11 | 1110 | ||
| AK148035.1 | 11 | 1123 | BAE28302.1 | |
| AK150785.1 | 11 | 1079 | BAE29849.1 | |
| AK151590.1 | 11 | 1135 | BAE30530.1 | |
| AK152296.1 | 12 | 1175 | BAE31105.1 | |
| AK153107.1 | 12 | 1175 | ||
| AK153144.1 | 11 | 1135 | BAE31755.1 | |
| AK165316.1 | 11 | 1095 | BAE38134.1 | |
| AK165944.1 | 11 | 1113 | BAE38476.1 | |
| AK167598.1 | 11 | 1068 | BAE39655.1 | |
| AK168400.1 | 11 | 1135 | ||
| AK169599.1 | 11 | 1106 | BAE41250.1 | |
| AK179745.1 | 2 | 138 | ||
| AK191496.1 | 2 | 160 | ||
| AK192557.1 | 2 | 221 | ||
| AK194307.1 | 2 | 111 | ||
| AK194339.1 | 2 | 222 | ||
| AK194722.1 | 2 | 119 | ||
| AK196134.1 | 2 | 109 | ||
| AK198762.1 | 2 | 108 | ||
| AK199865.1 | 2 | 221 | ||
| AK201262.1 | 2 | 221 | ||
| AK203011.1 | 2 | 221 | ||
| AK204019.1 | 2 | 221 | ||
| AK204900.1 | 2 | 116 | ||
| AK205283.1 | 1 | 77 | ||
| AK210768.1 | 2 | 117 | ||
| AK217077.1 | 2 | 221 | ||
| AK217357.1 | 2 | 131 | ||
| AK219356.1 | 2 | 221 | ||
| BC019119.1 | 11 | 1113 | AAH19119.1 |
More Information
Set Membership: |
|

Genomic Map