Assay Details
Target Gene Details
Entrez Gene ID: | 68275 |
Gene Name: | replication protein A1 |
Gene Aliases: |
5031405K23Rik, 70kDa, AA589576, AW557552, RF-A, RP-A, Rpa |
Location: |
Chr.11:75300259-75348383 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpa1 | NM_001164223.1 | 13 | 1382 | NP_001157695.1 |
| NM_026653.2 | 12 | 1319 | NP_080929.1 | |
| AK014298.1 | ||||
| AK019864.1 | 6 | 720 | ||
| AK035084.1 | 12 | 1247 | ||
| AK148035.1 | 12 | 1260 | BAE28302.1 | |
| AK150785.1 | 12 | 1216 | BAE29849.1 | |
| AK151590.1 | 12 | 1272 | BAE30530.1 | |
| AK152296.1 | 13 | 1312 | BAE31105.1 | |
| AK153107.1 | 13 | 1312 | ||
| AK153144.1 | 12 | 1272 | BAE31755.1 | |
| AK165316.1 | 12 | 1232 | BAE38134.1 | |
| AK165944.1 | 12 | 1250 | BAE38476.1 | |
| AK167598.1 | 12 | 1205 | BAE39655.1 | |
| AK168400.1 | 12 | 1272 | ||
| AK169599.1 | 12 | 1243 | BAE41250.1 | |
| AK179745.1 | 3 | 275 | ||
| AK191496.1 | 3 | 298 | ||
| AK192557.1 | 3 | 358 | ||
| AK194307.1 | 3 | 248 | ||
| AK194339.1 | 3 | 359 | ||
| AK194722.1 | 3 | 256 | ||
| AK196134.1 | 3 | 246 | ||
| AK198762.1 | 3 | 245 | ||
| AK199865.1 | ||||
| AK201262.1 | ||||
| AK203011.1 | ||||
| AK204019.1 | 3 | 358 | ||
| AK204900.1 | 3 | 253 | ||
| AK205283.1 | 2 | 214 | ||
| AK207399.1 | 1 | 58 | ||
| AK210768.1 | 3 | 254 | ||
| AK217077.1 | 3 | 358 | ||
| AK217357.1 | 3 | 268 | ||
| AK219356.1 | 3 | 357 | ||
| BC019119.1 | 12 | 1250 | AAH19119.1 |
More Information
Set Membership: |
|

Genomic Map