Assay Details
Target Gene Details
Entrez Gene ID: | 76547 |
Gene Name: | transmembrane protein 101 |
Gene Aliases: |
2610511E22Rik |
Location: |
Chr.11:102152547-102156407 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem101 | NM_029649.2 | NP_083925.2 | ||
| AK012106.1 | BAB28037.1 | |||
| AK041154.1 | BAC30841.1 | |||
| AK049378.1 | BAC33721.1 | |||
| AK140260.1 | BAE24302.1 | |||
| BC011109.1 | AAH11109.1 |

Genomic Map