Assay Details
Target Gene Details
Entrez Gene ID: | 71943 |
Gene Name: | target of myb1-like 1 (chicken) |
Gene Aliases: |
2310045L10Rik, C80573, Srcasm |
Location: |
Chr.11:90645690-90687661 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tom1l1 | NM_028011.2 | NP_082287.1 | ||
| AF395837.1 | AAK83377.1 | |||
| AJ505007.1 | CAD43734.1 | |||
| AK009826.1 | BAB26526.1 | |||
| AK033712.1 | BAC28440.1 | |||
| AK181527.1 | ||||
| BC004710.1 | AAH04710.1 |
More Information
Set Membership: |
|

Genomic Map