Assay Details
Target Gene Details
Entrez Gene ID: | 100504285 |
Gene Name: | abhydrolase domain containing 12B |
Gene Aliases: |
Gm804 |
Location: |
Chr.12:70154171-70183206 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd12b | NM_001195033.1 | NP_001181962.1 | ||
| AK136846.1 |
More Information
Set Membership: |
|

Genomic Map