Assay Details
Target Gene Details
Entrez Gene ID: | 171282 |
Gene Name: | acyl-CoA thioesterase 4 |
Gene Aliases: |
B430212I04Rik, PTE-Ib, Pte1b, Pte2b |
Location: |
Chr.12:84038379-84044723 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acot4 | NM_134247.3 | NP_599008.3 | ||
| AK046630.1 | BAC32814.1 | |||
| AK050420.1 | BAC34246.1 | |||
| AK080883.1 | BAC38060.1 | |||
| BC113185.1 |
More Information
Set Membership: |
|

Genomic Map