Assay Details
Target Gene Details
Entrez Gene ID: | 217700 |
Gene Name: | acyl-CoA thioesterase 6 |
Gene Aliases: |
4632408A20Rik, A330054B07, BE688602 |
Location: |
Chr.12:84100654-84109783 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acot6 | NM_172580.1 | NP_766168.1 | ||
| AY999300.1 | AAY45894.1 | |||
| BC139306.1 | ||||
| BC139328.1 |
More Information
Set Membership: |
|

Genomic Map