Assay Details
Target Gene Details
Entrez Gene ID: | 20787 |
Gene Name: | sterol regulatory element binding transcription factor 1 |
Gene Aliases: |
ADD-1, ADD1, D630008H06, SREBP-1, SREBP-1a, SREBP-1c, SREBP1, SREBP1c, bHLHd1 |
Location: |
Chr.11:60199089-60222686 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srebf1 | NM_011480.3 | 6 | 1170 | NP_035610.1 |
| AB017337.1 | 5 | 986 | BAA74795.1 | |
| AF374266.1 | 5 | 934 | AAK54762.1 | |
| AK052628.1 | 6 | 1171 | BAC35068.1 | |
| AK150052.1 | 6 | 1159 | BAE29268.1 | |
| AK154424.1 | 6 | 1138 | BAE32576.1 | |
| AK169607.1 | 6 | 1171 | BAE41256.1 | |
| BC006051.1 | 3 | 306 | AAH06051.1 | |
| BC056922.1 | 6 | 1152 | AAH56922.1 |

Genomic Map