Assay Details
Target Gene Details
Entrez Gene ID: | 216810 |
Gene Name: | target of myb1-like 2 (chicken) |
Gene Aliases: |
2900016I08Rik, A730055F12Rik, AU042072, Srebf1 |
Location: |
Chr.11:60223796-60352908 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tom1l2 | NM_001039092.3 | 3 | 276 | NP_001034181.1 |
| NM_001039093.1 | 3 | 276 | NP_001034182.1 | |
| NM_153080.2 | 3 | 276 | NP_694720.2 | |
| AF467887.1 | 3 | 256 | AAL77033.1 | |
| AK036788.1 | 3 | 252 | BAC29576.1 | |
| AK043095.1 | 3 | 256 | BAC31458.1 | |
| AK054531.1 | 3 | 244 | BAC35813.1 | |
| AK166040.1 | 3 | 276 | BAE38537.1 | |
| AK188563.1 | ||||
| BC062947.1 | 3 | 240 | ||
| BC127266.1 | 3 | 242 |

Genomic Map