Assay Details
Target Gene Details
Entrez Gene ID: | 74476 |
Gene Name: | RIKEN cDNA 4933439C10 gene |
Gene Aliases: |
2610028M21Rik |
Location: |
Chr.11:59505685-59511067 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933439C10Rik | NR_015585.2 | 1 | 130 | |
| AK017115.1 | 1 | 131 | ||
| AK163429.1 | 1 | 120 | ||
| AK169756.1 | 1 | 163 | BAE41348.1 |
Target Gene Details
Entrez Gene ID: | 268417 |
Gene Name: | zinc finger with KRAB and SCAN domains 17 |
Gene Aliases: |
Nizp1, Zfp496, Znf496 |
Location: |
Chr.11:59485520-59506640 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zkscan17 | NM_001130529.2 | NP_001124001.1 | ||
| NM_001291014.1 | NP_001277943.1 | |||
| NM_172941.4 | NP_766529.3 | |||
| AK051711.1 | BAC34730.1 | |||
| AK139346.1 | BAE23969.1 | |||
| AK155196.1 | BAE33110.1 | |||
| AK155490.1 | BAE33290.1 | |||
| AK157002.1 | BAE33928.1 | |||
| AK158448.1 | ||||
| AK170924.1 | BAE42117.1 | |||
| AK170989.1 | BAE42163.1 | |||
| AK171190.1 | BAE42300.1 | |||
| AK186703.1 | ||||
| AK190897.1 | ||||
| AK191778.1 | ||||
| AK209254.1 | ||||
| AY302242.1 | AAQ73562.1 | |||
| BC040205.1 | AAH40205.1 | |||
| BY753203.1 | ||||
| CB528232.1 |
More Information
Set Membership: |
|

Genomic Map